MilliporeSigma
  • Aminomethyltetrazoles as potential inhibitors of the γ-aminobutyric acid transporters mGAT1-mGAT4: synthesis and biological evaluation.
Aminomethyltetrazoles as potential inhibitors of the γ-aminobutyric acid transporters mGAT1-mGAT4: synthesis and biological evaluation.
Bioorganic & medicinal chemistry (2011-09-24)
Eva S Schaffert, Georg Höfner, Klaus T Wanner
ABSTRACT
1,5-Disubstituted and 5-monosubstituted aminomethyltetrazole derivatives derived from glycine were synthesized employing a TMSN(3)-modified variant of the Ugi reaction as a key step. All compounds were evaluated regarding their inhibitory potency and subtype selectivity at the four murine GABA transporter subtypes mGAT1-mGAT4. Though none of the 5-monosubstituted tetrazoles turned out to inhibit [(3)H]GABA uptake to a significant extent, the 1,5-disubstituted tetrazole derivatives displayed a distinct activity, especially at the GABA transport proteins mGAT2-mGAT4. Thus, a reasonable potent and selective inhibitor of mGAT3 was found. Additionally, two more compounds were identified as potent inhibitors of mGAT2. This is especially relevant, as up to date only few potent inhibitors of mGAT2 that do not affect mGAT1 are known.
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine 1 M solution