MilliporeSigma
  • Effects of Sildenafil Citrate and Heparin Treatments on Placental Cell Morphology in a Murine Model of Pregnancy Loss.
Effects of Sildenafil Citrate and Heparin Treatments on Placental Cell Morphology in a Murine Model of Pregnancy Loss.
Cells, tissues, organs (2016-03-16)
Rayana Leal Luna, Anne Gabrielle Vasconcelos, Ana Karolina Santana Nunes, Wilma Helena de Oliveira, Karla Patricia de Sousa Barbosa, Christina Alves Peixoto
ABSTRACT
Lipopolysaccharide (LPS) injections during pregnancy are well established as models for pregnancy complications, including fetal growth restriction (FGR), thrombophilia, preterm labor and abortion. Indeed, inflammation, as induced by LPS injection has been described as a pivotal factor in cases of miscarriage related to placental tissue damage. The phosphodiesterase-5 inhibitor sildenafil (Viagra®) is currently used to treat FGR cases in women, while low-molecular weight heparin (Fragmin®) is a standard treatment for recurrent miscarriage (RM). However, the pathways and cellular dynamics involved in RM are not completely understood. The aim of this study was to evaluate the protective effect of sildenafil and dalteparin in a mouse model of LPS-induced abortion. Histopathology, ultrastructural analysis and immunofluorescence for P-selectin were studied in two different placental cell types: trophoblast cells and labyrinth endothelial cells. Treatment with sildenafil either alone or in combination with heparin showed the best response against LPS-induced injury during pregnancy. In conclusion, our results support the use of these drugs as future therapeutic agents that may protect the placenta against inflammatory injury in RM events. Analyses of the ultrastructure and placental immunophysiology are important to understand the mechanism underlying RM. These findings may spark future studies and aid in the development of new therapies in cases of RM.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in sheep, affinity isolated antibody, buffered aqueous solution