MilliporeSigma
  • GSK973 Is an Inhibitor of the Second Bromodomains (BD2s) of the Bromodomain and Extra-Terminal (BET) Family.
GSK973 Is an Inhibitor of the Second Bromodomains (BD2s) of the Bromodomain and Extra-Terminal (BET) Family.
ACS medicinal chemistry letters (2020-08-25)
Alex Preston, Stephen J Atkinson, Paul Bamborough, Chun-Wa Chung, Laurie J Gordon, Paola Grandi, James R J Gray, Lee A Harrison, Antonia J Lewis, David Lugo, Cassie Messenger, Anne-Marie Michon, Darren J Mitchell, Rab K Prinjha, Inmaculada Rioja, Jon Seal, Simon Taylor, Pierre Thesmar, Ian D Wall, Robert J Watson, James M Woolven, Emmanuel H Demont
ABSTRACT
Pan-BET inhibitors have shown profound efficacy in a number of in vivo preclinical models and have entered the clinic in oncology trials where adverse events have been reported. These inhibitors interact equipotently with the eight bromodomains of the BET family of proteins. To better understand the contribution of each domain to their efficacy and to improve from their safety profile, selective inhibitors are required. This Letter discloses the profile of GSK973, a highly selective inhibitor of the second bromodomains of the BET proteins that has undergone extensive preclinical in vitro and in vivo characterization.
Sigma-Aldrich
GSK973, ≥98% (HPLC)
Sigma-Aldrich
GSK943, ≥98% (HPLC)