MilliporeSigma
Lignans from Trachelospermum jasminoides.
Planta medica (2005-01-29)
Xing-Qi Tan, Hai-Sheng Chen, Run-Hui Liu, Chang-Heng Tan, Cong-Li Xu, Wei-Dong Xuan, Wei-Dong Zhang
ABSTRACT
Seven lignans having a diarylhydroxybutyrolactone skeleton were isolated from the leaves and stems of Trachelospermum jasminoides (Lindl.) Lem. Their structures were elucidated to be nortrachelogenin 8' -O-beta-D-glucopyranoside (1), nortrachelogenin 5'- C- beta-D-glucopyranoside (2), trachelogenin amide (3), nortracheloside, trachelogenin, tracheloside, and trachelogenin 4'- O- beta-gentiobioside, respectively, on the basis of spectroscopic analyses. Lignans 1 - 3 were structurally identified to be new compounds, and 2 was a rare C-glucosyl-lignan.