MilliporeSigma
  • Alpha 2-adrenoceptor blockade prevents cardiac glycoside-evoked neurotransmitter release from sympathetic nerves in dog saphenous vein.
Alpha 2-adrenoceptor blockade prevents cardiac glycoside-evoked neurotransmitter release from sympathetic nerves in dog saphenous vein.
British journal of pharmacology (1987-09-01)
D A Powis
ABSTRACT
1 The effect of alpha-adrenoceptor antagonists upon neurotransmitter release evoked by cardiac glycosides from sympathetic nerve terminals has been investigated in dog saphenous vein. 2 In rings of saphenous vein preloaded with [3H]-noradrenaline, acetylstrophanthidin (ACS) caused a concentration-dependent efflux of 3H (EC50 ca. 4.4 microM) that was attenuated by phentolamine and yohimbine but not by prazosin. 3 In helical strips of saphenous vein superfused with ACS at EC50 the efflux of 3H-compounds in general, and of [3H]-noradrenaline in particular, occurred after a short delay and increased with time to a maximum reached at 75 min. Phentolamine and phenoxybenzamine, but not prazosin reduced the efflux of [3H]-noradrenaline and of total 3H-compounds throughout the time-course of the ACS-evoked effect. 4 In helical strips of saphenous vein the glycoside ouabain also caused an increase in [3H]-noradrenaline and in total 3H-efflux that was attenuated by phentolamine. 5 By contrast with the above, in bovine adrenal medullary chromaffin cells, which appear to have no functional alpha-adrenoceptors, ACS caused a small, but significant increase in 3H-efflux which was not prevented by phentolamine. 6 Phentolamine, at concentrations that attenuate markedly the ouabain- or ACS-evoked increase in 3H-efflux from dog saphenous vein, did not cause significant inhibition of cocaine-sensitive [3H]-noradrenaline uptake nor did it reduce the extent of the 3H-efflux evoked either by tyramine or by reduced extracellular Na+. These findings imply that phentolamine does not affect ACS-evoked neurotransmitter release by an action on the catecholamine uptake mechanism. 7. It is concluded that the cardiac glycoside-evoked increase in neurotransmitter release from noradrenergic nerve terminals ofdog saphenous vein is modulated by a mechanism that involves an alpha2- adrenoceptor.
Sigma-Aldrich
Strophanthidin