MilliporeSigma
  • Gag- and Nef-specific CD4+ T cells recognize and inhibit SIV replication in infected macrophages early after infection.
Gag- and Nef-specific CD4+ T cells recognize and inhibit SIV replication in infected macrophages early after infection.
Proceedings of the National Academy of Sciences of the United States of America (2009-05-30)
Jonah B Sacha, Juan P Giraldo-Vela, Matthew B Buechler, Mauricio A Martins, Nicholas J Maness, Chungwon Chung, Lyle T Wallace, Enrique J León, Thomas C Friedrich, Nancy A Wilson, Atsunobu Hiraoka, David I Watkins
ABSTRACT
The precise immunological role played by CD4(+) T cells in retroviral infections is poorly defined. Here, we describe a new function of these cells, the elimination of retrovirus-infected macrophages. After experimental CD8(+) cell depletion, elite controlling macaques with set-point viral loads < or = 500 viral RNA copies/mL mounted robust Gag- and Nef-specific CD4(+) T cell responses during reestablishment of control with > or = 54% of all virus-specific CD4(+) T cells targeting these 2 proteins. Ex vivo, these simian immunodeficiency virus (SIV)-specific CD4(+) T cells neither recognized nor suppressed viral replication in SIV-infected CD4(+) T cells. In contrast, they recognized SIV-infected macrophages as early as 2 h postinfection because of presentation of epitopes derived from virion-associated Gag and Nef proteins. Furthermore, virus-specific CD4(+) T cells displayed direct effector function and eliminated SIV-infected macrophages. These results suggest that retrovirus-specific CD4(+) T cells may contribute directly to elite control by inhibiting viral replication in macrophages.
Sigma-Aldrich
M-CSF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
M-CSF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
M-CSF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture