MilliporeSigma
  • Initiation codon mutation of the tyrosinase gene as a cause of human albinism.
Initiation codon mutation of the tyrosinase gene as a cause of human albinism.
Clinica chimica acta; international journal of clinical chemistry (1994-06-01)
L H Breimer, A F Winder, B Jay, M Jay
ABSTRACT
Direct DNA sequence determination of PCR amplified exons of the tyrosinase gene of three British patients suffering from tyrosinase negative oculocutaneous albinism has revealed three new missense point mutations: (1) an adenine to guanine transition at codon 1 changes the initiating methionine codon into a valine codon thereby abolishing translation; (2) a thymine to cytosine transition at codon 370 changes a methionine to a threonine residue; (3) a cytosine to thymine transition at codon 367 changes a histidine to a tyrosine residue. A codon 402 change previously considered a polymorphism is assigned a pathological role.
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid