MilliporeSigma
  • Enzymatic preparation of maltohexaose, maltoheptaose, and maltooctaose by the preferential cyclomaltooligosaccharide (cyclodextrin) ring-opening reaction of Pyrococcus furiosus thermostable amylase.
Enzymatic preparation of maltohexaose, maltoheptaose, and maltooctaose by the preferential cyclomaltooligosaccharide (cyclodextrin) ring-opening reaction of Pyrococcus furiosus thermostable amylase.
Carbohydrate research (2005-12-21)
Sung-Jae Yang, Hee-Seob Lee, Jung-Woo Kim, Myoung-Hee Lee, Joong-Hyuck Auh, Byong-Hoon Lee, Kwan-Hwa Park
ABSTRACT
Specific-length maltooligosaccharides, particularly maltohexaose, maltoheptaose, and maltooctaose, were prepared from cyclomaltooligosaccharides (cyclodextrins, CDs) by the preferential cyclodextrin ring-opening reaction of an amylolytic enzyme from Pyrococcus furiosus. The enzyme primarily produces maltohexaose, maltoheptaose, and maltooctaose by hydrolyzing alpha-, beta-, and gamma-CD, respectively. This study aims to develop a high-efficiency synthesis of specific maltooligosaccharides at high-purity. [formula: see text]
Sigma-Aldrich
Maltohexaose, ≥65% (HPLC)
Sigma-Aldrich
Maltoheptaose, ≥60% (HPLC)