MilliporeSigma
  • Silica bonded S-sulfonic acid: a recyclable catalyst for the synthesis of quinoxalines at room temperature.
Silica bonded S-sulfonic acid: a recyclable catalyst for the synthesis of quinoxalines at room temperature.
Molecules (Basel, Switzerland) (2009-05-28)
Khodabakhsh Niknam, Dariush Saberi, Maleki Mohagheghnejad
ABSTRACT
The reaction of 3-mercaptopropylsilica (MPS) and chlorosulfonic acid in chloroform afforded silica bonded S-sulfonic acid (SBSSA), which was used as a catalyst for the room temperature synthesis of quinoxaline derivatives from 1,2-diamino compounds and 1,2-dicarbonyl compounds. The catalyst could be recycled and reused several times without any loss of efficiency.
Sigma-Aldrich
Chlorosulfonic acid, 99%