MilliporeSigma
  • Liquid antisolvent preparation of amorphous cefuroxime axetil nanoparticles in a tube-in-tube microchannel reactor.
Liquid antisolvent preparation of amorphous cefuroxime axetil nanoparticles in a tube-in-tube microchannel reactor.
International journal of pharmaceutics (2010-05-25)
Wen-Zhen Zhu, Jie-Xin Wang, Lei Shao, Hai-xia Zhang, Qian-xia Zhang, Jian-Feng Chen
ABSTRACT
This article presents the preparation of nanoparticles of amorphous cefuroxime axetil (CFA) in a microporous tube-in-tube microchannel reactor (MTMCR). The experimental results indicated that CFA particle with a tunable size of 400-1400 nm could be achieved under a high throughput in the range of 1.5-6L/min. The average particle size decreased with increasing overall volumetric flow rate and decreasing CFA concentration, micropore size, and annular channel width. The produced CFA nanoparticles were characterized by SEM, XRD, FT-IR, DSC and a dissolution test, which indicated that the nanosized CFA was amorphous and exhibited higher dissolution rate compared to the raw CFA. The MTMCR might offer a general and facile pathway for mass production of the nanoparticles of hydrophobic pharmaceuticals thanks to its high throughput capacity and excellent micromixing performance.
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Supelco
Diisopropyl ether, analytical standard
Sigma-Aldrich
Diisopropyl ether, ReagentPlus®, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Diisopropyl ether, contains either BHT or hydroquinone as stabilizer, ACS reagent, ≥99.0%
Sigma-Aldrich
Diisopropyl ether, puriss. p.a., ≥98.5% (GC)