MilliporeSigma
  • Synthesis and evaluation of a set of para-substituted 4-phenylpiperidines and 4-phenylpiperazines as monoamine oxidase (MAO) inhibitors.
Synthesis and evaluation of a set of para-substituted 4-phenylpiperidines and 4-phenylpiperazines as monoamine oxidase (MAO) inhibitors.
Journal of medicinal chemistry (2012-03-06)
Fredrik Pettersson, Peder Svensson, Susanna Waters, Nicholas Waters, Clas Sonesson
ABSTRACT
A series of para-substituted 4-phenylpiperidines/piperazines have been synthesized and their affinity to recombinant rat cerebral cortex monoamine oxidases A (MAO A) and B (MAO B) determined. Para-substituents with low dipole moment increased the affinity to MAO A, whereas groups with high dipole moment yielded compounds with no or weak affinity. In contrast, the properties affecting MAO B affinity were the polarity and bulk of the para-substituent, with large hydrophobic substituents producing compounds with high MAO B affinity. In addition, these compounds were tested in freely moving rats and the effect on the post-mortem neurochemistry was measured. A linear correlation was demonstrated between the affinity for MAO A, but not MAO B, and the levels of 3,4-dihydroxyphenylacetic acid (DOPAC) and 3-methoxytyramine (3-MT) in the striatum.
Sigma-Aldrich
3-Methoxytyramine hydrochloride, ≥95.5%, crystalline
Sigma-Aldrich
3-Methoxytyramine hydrochloride, 99% (AT)