MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Cavitation-induced radical-chain oxidation of valeric aldehyde.
Ultrasonics sonochemistry (2012-03-06)
Ulrich Neuenschwander, Jürg Neuenschwander, Ive Hermans
ABSTRACT
The application of high-amplitude ultrasound to liquids triggers cavitation. By the collapse of the thereby appearing vacuum cavities, high temperatures can be reached in a transient manner. The high temperatures in these hot-spots can lead to homolytic scission of chemical bonds. The thereby generated radicals are usually utilized in aqueous systems for the degeneration of organic pollutants. In this contribution, we demonstrate that the radicals can also be used for synthetic purposes: under an oxygen atmosphere, they trigger the oxidation of an aldehyde substrate.
Sigma-Aldrich
Valeraldehyde, ≥97%, FG
Sigma-Aldrich
Valeraldehyde, 97%