MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Methiocarb poisoning of a horse in Australia.
Australian veterinary journal (2012-05-29)
B M Kaye, C R B Elliott, S L Jalim
ABSTRACT
Snail bait poisoning is rare in horses. Cases have been reported, but clinical signs and subsequent prognostic indicators have been poorly documented and must be extrapolated from cases in companion animals. We describe in detail the poisoning of a horse that consumed a lethal dose of the carbamate, methiocarb. There are currently no guidelines for treating equine methiocarb toxicoses, but the principles of management are based on supportive therapy.
Product Description
Supelco
Methiocarb, PESTANAL®, analytical standard