MilliporeSigma
  • [Preparation and in vitro release characteristics of pulsed-release tablets of Apocynum venetum].
[Preparation and in vitro release characteristics of pulsed-release tablets of Apocynum venetum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-07-12)
Huasheng Yang, Fugui Xie, Yuewen Yang, Yongming Luo
ABSTRACT
Using Apocynum venetum as a model drug to prepare pulsed-release tablets based on diffusion, swelling, osmotic pressure mechanism and to evaluate the release characteristics. The pulsatile release tablets were prepared by film coating methods using HPMC E5 and Eudragit The effect of formulation on pulsatile release of A. venetum was investigated. The pulsed-release tablet was prepared by a swelling layer coating which contains HPMC E5 and a controlled-release membrane containning Eudragit. The delayed release time of the tablets was (5.0 +/- 0.5) h. The pulsatile release characteristics of A. venetum pulsatile release tablets were confirmed in vitro.
Sigma-Aldrich
Poly(methyl methacrylate-co-methacrylic acid), average Mw ~34,000 by GPC, average Mn ~15,000 by GPC