MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Plant cell biology: the ABC of monolignol transport.
Current biology : CB (2012-07-14)
Richard Sibout, Herman Höfte
ABSTRACT
Lignins are complex aromatic heteropolymers that reinforce the cell walls of terrestrial plants. A new study identifies an ATP-binding cassette ABC transporter that pumps a monolignol lignin precursor across the plasma membrane.
Sigma-Aldrich
Coniferyl alcohol, 98%