MilliporeSigma
  • Effectiveness of dry eye therapy under conditions of environmental stress.
Effectiveness of dry eye therapy under conditions of environmental stress.
Current eye research (2013-01-09)
Alan Tomlinson, Louise C Madden, Peter A Simmons
ABSTRACT
Dry eye is often characterized by increased tear evaporation due to poor tear film quality, especially of the lipid component of the tear film. Using an environmental chamber to induce environmental stress, this study compared the effect of three lubricant eye drops on various aspects of tear physiology in a crossover design (evaporation was the principal outcome measure). Three eye drop formulas were tested: 0.5% carmellose sodium (Drop C), 0.5% carmellose sodium with added lipid (Drop C-L) and 1.0% glycerine with added lipid (Drop G-L). Nineteen control and 18 dry eye subjects used each product for 2 weeks, three times per day, in a random order, with a minimum 1-week washout between treatment periods. Tear evaporation, break up time, osmolarity, tear structure (by interferometry) and patient symptoms were assessed with the subjects adapted for 10 min in an environmental chamber controlled at 20% relative humidity and 22 °C. The treatment effects were analyzed using general linear model repeated measures analyses of variance. In dry eye subjects, evaporation, break up time, osmolarity and symptoms improved for all formulas (p < 0.05). Normal subjects showed some improvements: evaporation with C-L, osmolarity with C and symptoms with C-L and G-L. Change in evaporation was greater for both C-L and G-L versus C (p < 0.05), and there was a trend for C-L to reduce evaporation more than G-L (p < 0.11). There were no significant treatment effects on tear film structure. Overall, the eye drop formula containing both carmellose sodium and lipid (C-L) produced a greater treatment effect on tear evaporation than the other formulations containing only one of these ingredients. This study also demonstrates the utility of a controlled environmental chamber in showing the difference in performance between dry eye treatments.
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity