MilliporeSigma
Use of glycopyrrolate and atropine in acute organophosphorus poisoning.
Human & experimental toxicology (1990-03-01)
J A Tracey, H Gallagher
ABSTRACT
Two cases of organophosphorus poisoning are reported. Both were treated with a combination of atropine and glycopyrrolate as well as benzodiazepine and pralidoxime. The advantages of glycopyrrolate over atropine are discussed.