MilliporeSigma
  • Aedes aegypti mosquitoes exhibit decreased repellency by DEET following previous exposure.
Aedes aegypti mosquitoes exhibit decreased repellency by DEET following previous exposure.
PloS one (2013-02-26)
Nina M Stanczyk, John F Y Brookfield, Linda M Field, James G Logan
ABSTRACT
DEET (N,N-Diethyl-m-toluamide) is one of the most widely used mosquito repellents. Although DEET has been shown to be extremely effective, recent studies have revealed that certain individual insects are unaffected by its presence. A genetic basis for this has been shown in Aedes aegypti mosquitoes and the fruit fly Drosophila melanogaster, but, for the triatomine bug, Rhodnius prolixus, a decrease in response to DEET occurred shortly after previous exposure, indicating that non-genetic factors may also be involved in DEET "insensitivity". In this study, we examined host-seeking behaviour and electrophysiological responses of A. aegypti after pre-exposure to DEET. We found that three hours after pre-exposure the mosquitoes showed behavioural insensitivity, and electroantennography revealed this correlated with the olfactory receptor neurons responding less to DEET. The change in behaviour as a result of pre-exposure to DEET has implications for the use of repellents and the ability of mosquitoes to overcome them.
Sigma-Aldrich
N,N-Diethyl-3-methylbenzamide, 97%
Supelco
DEET, PESTANAL®, analytical standard