MilliporeSigma
  • Rhodium(III)-catalyzed direct selective C(5)-H oxidative annulations of 2-substituted imidazoles and alkynes by double C-H activation.
Rhodium(III)-catalyzed direct selective C(5)-H oxidative annulations of 2-substituted imidazoles and alkynes by double C-H activation.
Organic letters (2013-03-30)
Ji-Rong Huang, Qian-Ru Zhang, Chuan-Hua Qu, Xun-Hao Sun, Lin Dong, Ying-Chun Chen
ABSTRACT
Double C-H activations of C(5)-H and Csp(2)-H of 2-substituted N-vinyl- or arylimidazoles were realized without heteroatom-directing assistance by rhodium(III) catalyst. A subsequent oxidative annulation reaction with alkynes efficiently produced aza-fused heterocycles with high molecular complexity in low to excellent yields.
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis