MilliporeSigma
  • Electrophoretic heterogeneity limits the utility of streptavidin-β-galactosidase as a probe in free zone capillary electrophoresis separations.
Electrophoretic heterogeneity limits the utility of streptavidin-β-galactosidase as a probe in free zone capillary electrophoresis separations.
The protein journal (2013-01-18)
Douglas B Craig, Anna Henderson
ABSTRACT
Single molecule assays were performed on streptavidin-β-galactosidase using a capillary electrophoresis-based protocol in order to assess the suitability of single molecule β-galactosidase assays for adaptation to the detection of single copies of target DNA. The conjugate was found to have a heterogeneous catalytic rate, showing an average rate of 44,000 ± 24,000 min(-1), which is similar to that of the unmodified enzyme. Electrophoretic mobility was also measured on individual molecules and determined to be -1.32 × 10(-4) ± 0.19 × 10(-4) cm(2)V(-1)s(-1). The variance in mobility was several times that reported for the unmodified enzyme. The electrophoretic heterogeneity was found to result in the formation of a broad window of peaks in the resultant electropherograms of free zone separations of small plugs of streptavidin-β-galactosidase. This range of mobilities largely overlapped with that of the conjugate bound to primer and plasmid containing a target DNA sequence. This overlap suggests that the separation of free conjugate from that bound to target DNA, which is a requirement for application of the single enzyme molecule assay to the detection of target DNA sequences, is not plausible using free zone capillary electrophoresis.
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥13 units/mg protein
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, affinity purified, lyophilized from 10 mM potassium phosphate, ≥13 U/mg protein
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, recombinant, expressed in E. coli, lyophilized powder