MilliporeSigma
  • 1-Nitro-2-phenylethane, the main constituent of the essential oil of Aniba canelilla, elicits a vago-vagal bradycardiac and depressor reflex in normotensive rats.
1-Nitro-2-phenylethane, the main constituent of the essential oil of Aniba canelilla, elicits a vago-vagal bradycardiac and depressor reflex in normotensive rats.
European journal of pharmacology (2010-04-22)
Rodrigo José Bezerra de Siqueira, Francisco Igor Bulcão Macedo, Leylliane de Fátima Leal Interaminense, Gloria Pinto Duarte, Pedro Jorge Caldas Magalhães, Teresinha Silva Brito, Joyce Kelly Rosário da Silva, José Guilherme Soares Maia, Pergentino José da Cunha Sousa, José Henrique Leal-Cardoso, Saad Lahlou
ABSTRACT
Previously, it was shown that intravenous (i.v.) treatment with the essential oil of Aniba canelilla (EOAC) elicited a hypotensive response that is due to active vascular relaxation rather than to the withdrawal of sympathetic tone. The present study investigated mechanisms underlying the cardiovascular responses to 1-nitro-2-phenylethane, the main constituent of the EOAC. In pentobarbital-anesthetized normotensive rats, 1-nitro-2-phenylethane (1-10mg/kg, i.v.) elicited dose-dependent hypotensive and bradycardiac effects which were characterized in two periods (phases 1 and 2). The first rapid component (phase 1) evoked by 1-nitro-2-phenylethane (10mg/kg) was fully abolished by bilateral vagotomy, perineural treatment of both cervical vagus nerves with capsaicin (250 microg/ml) and was absent after left ventricle injection. However, pretreatment with capsazepine (1mg/kg, i.v.) or ondansetron (30 microg/kg, i.v.) did not alter phase 1 of the cardiovascular responses to 1-nitro-2-phenylethane (10mg/kg, i.v.). In conscious rats, 1-nitro-2-phenylethane (1-10mg/kg, i.v.) evoked rapid hypotensive and bradycardiac (phase 1) effects that were fully abolished by methylatropine (1mg/kg, i.v.). It is concluded that 1-nitro-2-phenylethane induces a vago-vagal bradycardiac and depressor reflex (phase 1) that apparently results from the stimulation of vagal pulmonary rather than cardiac C-fiber afferents. The transduction mechanism of the 1-nitro-2-phenylethane excitation of C-fiber endings is not fully understood and does not appear to involve activation of either Vanilloid TPRV(1) or 5-HT(3) receptors. The phase 2 hypotensive response to 1-nitro-2-phenylethane seems to result, at least in part, from a direct vasodilatory effect since 1-nitro-2-phenylethane (1-300 microg/ml) induced a concentration-dependent reduction of phenylephrine-induced contraction in rat endothelium-containing aorta preparations.
Product Description
Methylatropine bromide, European Pharmacopoeia (EP) Reference Standard