MilliporeSigma
  • Validated LC-MS/MS assay for the quantitative determination of nalbuphine in human plasma and its application to a pharmacokinetic study.
Validated LC-MS/MS assay for the quantitative determination of nalbuphine in human plasma and its application to a pharmacokinetic study.
Biomedical chromatography : BMC (2011-02-22)
Li-Jing Cai, Jun Zhang, Xiu-Mei Wang, R H Zhu, Jian Yang, Qi-Zhi Zhang, W X Peng
ABSTRACT
A solid-phase extraction-liquid chromatographic-tandem mass spectrometry method for the determination of nalbuphine concentrations in human plasma has been developed. Samples (1 mL) were extracted using a Strata™-X solid phase extraction cartridges. Chromatographic separation of nalbuphine and naloxone (internal standard) was achieved on a Phenomenex Kinetex PFP (2.6 μm, 100 A, 100 × 2.1 mm) column using a mobile phase consisting of 0.1% formic acid, 15 mM ammonium acetate in deionized water and acetonitrile (60:40, v/v). The flow rate was 0.3 mL/min and the total run time was 2 min. Detection of the analytes was achieved using positive ion electrospray ionization via multiple reactions monitoring mode. The mass transitions were m/z 358 → 340 for nalbuphine and m/z 328 → 310 for naloxone. The assay was linear over the concentration range 0.50-500.00 ng/mL, with correlation coefficients ≥0.995. The lower limit of quantitation was set at 0.5 ng/mL plasma based on an average signal-to-noise ratio of 44.79. The intra- and inter-day precision was less than 8.07% in terms of relative standard deviation and accuracy ranged from 94.97 to 106.29% at all quality control levels. The method was applied successfully to determine nalbuphine concentrations in human plasma samples obtained from subjects receiving intravenous administration of nalbuphine. The method is rapid, sensitive, selective and directly applicable to human pharmacokinetic studies involving nalbuphine.
Product Description
Supelco
Nalbuphine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®