MilliporeSigma
  • Ginsenoside-Rd potentiates apoptosis induced by hydrogen peroxide in basilar artery smooth muscle cells through the mitochondrial pathway.
Ginsenoside-Rd potentiates apoptosis induced by hydrogen peroxide in basilar artery smooth muscle cells through the mitochondrial pathway.
Apoptosis : an international journal on programmed cell death (2011-11-15)
Shi-Yang Li, Xiao-Guang Wang, Ming-Ming Ma, Yun Liu, Yan-Hua Du, Xiao-Fei Lv, Jia-Guo Zhou, Yong-Bo Tang, Yong-Yuan Guan
ABSTRACT
Our previous studies showed that ginsenoside-Rd, a purified component from Panax notoginseng, inhibited cell proliferation and reversed basilar artery remodeling. The aim of this study was to investigate whether ginsenoside- Rd influences H(2)O(2)-induced apoptosis in basilar artery smooth muscle cells (BASMCs). The results showed that ginsenoside-Rd significantly potentiated H(2)O(2)-induced cell death and cell apoptosis. This resulted in a concentration-dependent reduction of the cell viability. Ginsenoside-Rd further increased cytochrome C release and caspase-9/caspase-3 activations, and reduced the stability of mitochondrial membrane potential (MMP) and the ratio of Bcl-2/Bax. Cyclosporine A, an inhibitor of mitochondrial-permeability transition, inhibited alteration of mitochondrial permeability induced by H(2)O(2) and reversed the effect of ginsenoside-Rd on MMP. Our data strongly suggest that ginsenoside-Rd potentiated H(2)O(2)-induced apoptosis of BASMCs through the mitochondria-dependent pathway.