MilliporeSigma
  • Design and synthesis of novel stiripentol analogues as potential anticonvulsants.
Design and synthesis of novel stiripentol analogues as potential anticonvulsants.
European journal of medicinal chemistry (2011-11-29)
Mohamed N Aboul-Enein, Aida A El-Azzouny, Mohamed I Attia, Yousreya A Maklad, Kamilia M Amin, Mohamed Abdel-Rehim, Mohammed F El-Behairy
ABSTRACT
A series of stiripentol (STP) analogues namely, 2-[(1E)-1-(1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-ylidene]-N-(aryl/H)hydrazinecarboxamides 7a-h, (±)-(5RS)-N-(aryl/H)-(1,3-benzodioxol-5-yl)-3-tert-butyl-4,5-dihydro-1H-pyrazole-1-carboxamides (±)-8a-h, and (±)-[(5RS)-(1,3-benzodioxol-5-yl)-3-tert-butyl-4,5-dihydro-1H-pyrazol-1-yl](aryl)methanones (±)-13a-f was synthesized by adopting appropriate synthetic routes and was pharmacologically evaluated in the preliminary anticonvulsant screens. The selected bioactive new chemical entities were subjected to ED(50) determination and neurotoxicity evaluation. The most active congeners are 7h in MES screen and (±)-13b in scPTZ screen which displayed ED(50) values of 87 and 110 mg/kg, respectively, as compared to that of STP (ED(50) = 277.7 and 115 mg/kg in MES and scPTZ, respectively).
Sigma-Aldrich
Stiripentol, ≥98% (HPLC)