MilliporeSigma
  • Novel nanoporous bioceramic spheres for drug delivery application: a preliminary in vitro investigation.
Novel nanoporous bioceramic spheres for drug delivery application: a preliminary in vitro investigation.
Oral surgery, oral medicine, oral pathology and oral radiology (2013-03-05)
Deepti Simon, Suvy Manuel, Harikrishna Varma
ABSTRACT
Bioceramics and their composites have found myriad applications in medicine as superlative osteoalloplasts. Their potential to function as a biocompatible resorbable drug delivery system is being explored. The present study is a preliminary investigation into the efficacy of these indigenously developed nanoporous materials as vehicles for therapeutic agents. An in vitro experiment was conducted with the goal of assessing this material and comparing it with a commercially available gentamicin-loaded polymethylmethacrylate cement. The drug-eluting characteristics of gentamicin bone cement and indigenously designed nanoporous bioceramic granules were analyzed spectrophotometrically and compared. Regression analysis was done. The first 5 days saw the elute from both samples containing drug concentrations >100 μg/g. Both samples exhibit a high initial-burst release, which is ideal for prophylactic purposes. Drug eluent levels for both materials were >100 μg/g, which is sufficient for bactericidal activity.
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 150,000 (average), 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed