MilliporeSigma
  • A phantom using titanium and Landolt rings for image quality evaluation in mammography.
A phantom using titanium and Landolt rings for image quality evaluation in mammography.
Physics in medicine and biology (2013-03-27)
Hugo de las Heras, Felix Schöfer, Britta Tiller, Margarita Chevalier, Georg Zwettler, Friedrich Semturs
ABSTRACT
A phantom for image quality evaluation of digital mammography systems is presented and compared to the most widely used phantoms in Europe and the US. The phantom contains objects for subjective detection of Landolt rings (four-alternative, forced-choice task) and for objective calculation of signal-difference-to-noise ratios (SDNR), both in a titanium background within a 12-step wedge. Evaluating phantom images corresponding to exposures between 15 and 160 mAs (average glandular dose between 0.2 and 2 mGy), the resulting scores were compared to the scores obtained following the European EPQC and American College of Radiology (ACR) protocols. Scores of the Landolt test equal to 19 and 8.5 and SDNR equal to 20 and 11 were found to be equivalent to the acceptable limiting values suggested by EPQC and ACR. In addition, the Landolt and SDNR tests were shown to take into account the anatomical variations in thickness and tissue density within the breast. The simplified evaluation method presented was shown to be a sensitive, efficient and reliable alternative for image quality evaluation of mammography systems.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic