MilliporeSigma
  • Measurement of vitamin A, vitamin E, selenium, and L-lactate in dogs with and without osteoarthritis secondary to ruptured cranial cruciate ligament.
Measurement of vitamin A, vitamin E, selenium, and L-lactate in dogs with and without osteoarthritis secondary to ruptured cranial cruciate ligament.
The Canadian veterinary journal = La revue veterinaire canadienne (2013-06-05)
Alexander de Oliveira El-Warrak, Mouhamed Rouma, Audrey Amoroso, Soren R Boysen, Younés Chorfi
ABSTRACT
This study compared vitamin A, vitamin E, selenium (Se), and L-lactate in blood and synovial fluid in 2 groups of 6 dogs; a control group (without OA) and an osteoarthritic group with spontaneous cranial cruciate ligament rupture and OA. Concentrations of vitamin E were significantly higher in serum than in synovial fluid in both OA (P = 0.006) and control (P = 0.0008) groups. Vitamin E concentration in synovial fluid was significantly higher in the OA group than in the control group (P = 0.009). Concentrations of Se were significantly higher in serum than in synovial fluid in both OA (P = 0.003) and control (P = 0.0006) groups. There were no significant differences in levels of Se, vitamin A, and L-lactate between the 2 groups. This is the first study to show an increased concentration of vitamin E in the synovial fluid of dogs with OA compared with dogs that did not have OA.
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis