MilliporeSigma
  • Platelet-rich plasma à la carte: commentary on an article by Satoshi Terada, MD, et al.: "use of an antifibrotic agent improves the effect of platelet-rich plasma on muscle healing after injury".
Platelet-rich plasma à la carte: commentary on an article by Satoshi Terada, MD, et al.: "use of an antifibrotic agent improves the effect of platelet-rich plasma on muscle healing after injury".
The Journal of bone and joint surgery. American volume (2013-06-20)
Christopher H Evans
Product Description
Losartan potassium, European Pharmacopoeia (EP) Reference Standard
Supelco
Losartan potassium