MilliporeSigma
  • Presence of immunoreactive alpha-bag cell peptide[1-8] in bag cell neurons of Aplysia suggests novel carboxypeptidase processing of neuropeptides.
Presence of immunoreactive alpha-bag cell peptide[1-8] in bag cell neurons of Aplysia suggests novel carboxypeptidase processing of neuropeptides.
Neuropeptides (1987-10-01)
S M Pulst, B S Rothman, E Mayeri
ABSTRACT
alpha-bag cell peptide (alpha BCP) is a putative neurotransmitter released from bag cell neurons of the marine mollusc Aplysia. alpha BCP is present in bag cell extracts and releasate from bag cells in two neuroactive forms: alpha BCP[1-9] and alpha BCP[1-8]. alpha BCP[1-8] is 30 times as potent as [1-9] in inhibiting target neurons, suggesting that both forms of the peptide serve as neurotransmitters. However, biochemical and molecular genetic data suggest that only alpha BCP[1-9] is originally cleaved directly from a larger precursor protein and that generation of alpha BCP[1-8] would require an unusual C-terminal leucine cleavage of alpha BCP[1-9]. To further ascertain which forms of alpha BCP are normally present in bag cells, we generated highly specific antisera to each peptide. We found intense immunostaining for both peptides in bag cell somata and nerve terminals. Moreover, both forms were stable in bag cell extract for at least 1 hr, which suggests that proteolysis in the extracts had been effectively inhibited. These results suggest that both alpha BCP[1-8] and [1-9] are normally present in bag cell somata and terminals and that a small amount of alpha BCP[1-9] is processed to alpha BCP[1-8] in vesicles before release. The results support the interpretation that the activity of an intravesicular carboxypeptidase generates alpha BCP[1-8] and thereby regulates the amount of inhibitory activity released during a bag cell discharge.