MilliporeSigma
Acetaldehyde acutely impairs canine testicular testosterone secretion.
European journal of pharmacology (1981-04-09)
T W Boyden, M A Silvert, R W Pamenter
ABSTRACT
Utilizing a method for perfusion of the in vivo isolated canine testis we have examined basal and human chorionic gonadotropin (hGC)-stimulated testosterone production of testes acutely exposed to ethanol and acetaldehyde. Ethanol infused at concentrations of 0.2 g/dl to 0.6 g/dl did not alter basal of hCG-stimulated testicular testosterone production of one testis when compared to the saline-infused control testis of the same animal. However, acetaldehyde infused at a concentration of 0.2 mg/dkl, a level similar to that found in humans drinking moderate amounts of ethanol, significantly impaired hCG-stimulated testicular testosterone production. It is concluded that acetaldehyde acutely impairs hCG-stimulated testicular testosterone secretion by a direct effect on the testis, but ethanol does not.
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF