MilliporeSigma
  • Amino acid sequence of proteinase K from the mold Tritirachium album Limber: Proteinase K ? a subtilisin-related enzyme with disulfide bonds.
Amino acid sequence of proteinase K from the mold <I>Tritirachium album</I> Limber: Proteinase K ? a subtilisin-related enzyme with disulfide bonds.
Febs Letters (1986)
Jany KD, et al.