MilliporeSigma
  • Catalytic, asymmetric, interrupted Feist-Bénary reactions of α-tosyloxyacetophenones.
Catalytic, asymmetric, interrupted Feist-Bénary reactions of α-tosyloxyacetophenones.
Organic letters (2011-11-08)
Michael A Calter, Alexander Korotkov
ABSTRACT
A new variant of the Interrupted Feist-Bénary (IFB) reaction uses α-tosyloxyacetophenones as electrophiles and proceeds in good yields and excellent enantioselectivities.