MilliporeSigma
  • Enzyme-resistant starch. III. The quality of straight-dough bread containing varying levels of enzyme-resistant starch.
Enzyme-resistant starch. III. The quality of straight-dough bread containing varying levels of enzyme-resistant starch.
Cereal Chem. (1994)
Eerlingen, R. C., et al.