MilliporeSigma
  • 14-3-3ε and NAV2 interact to regulate neurite outgrowth and axon elongation.
14-3-3ε and NAV2 interact to regulate neurite outgrowth and axon elongation.
Archives of biochemistry and biophysics (2013-10-29)
Mark A Marzinke, Terri Mavencamp, Joseph Duratinsky, Margaret Clagett-Dame
ABSTRACT
Neuron navigator 2 (NAV2) is required for all-trans retinoic acid (atRA) to induce neurite outgrowth in human neuroblastoma cells. Further, ectopic overexpression of full-length human NAV2 rescues an axonal elongation defect in the Caenorhabditis elegans unc-53 (NAV2 ortholog) mutant. Using a region of NAV2 that independently associates with the cytoskeleton as bait in a yeast-two-hybrid screen, 14-3-3ε was identified as a novel NAV2 interacting partner. Amino acids 761-960 of NAV2 are sufficient to confer a positive interaction with 14-3-3ε as evidenced by a two-hybrid screen and co-immunoprecipitation assay. Knockdown of 14-3-3ε leads to a decrease in atRA-mediated neurite outgrowth, similar to the elongation defects observed when NAV2 is depleted or mutated. Likewise, posterior lateral microtubule (PLM) defects in C. elegans fed unc-53 RNAi are similar to those fed ftt-2 (14-3-3 homolog) RNAi. The discovery of an interaction between NAV2 and 14-3-3ε could provide insight into the mechanism by which NAV2 participates in promoting cell migration and neuronal elongation.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Tretinoin, European Pharmacopoeia (EP) Reference Standard
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2