MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of growth rate on plasmid DNA production and metabolic performance of engineered Escherichia coli strains.
Effect of growth rate on plasmid DNA production and metabolic performance of engineered Escherichia coli strains.
Journal of bioscience and bioengineering (2013-09-10)
Martin Wunderlich, Hilal Taymaz-Nikerel, Guillermo Gosset, Octavio T Ramírez, Alvaro R Lara
ABSTRACT
Two engineered Escherichia coli strains, designated VH33 and VH34, were compared to their parent strain W3110 in chemostat mode during plasmid DNA (pDNA) production. In strain VH33 the glucose uptake system was modified with the aim of reducing overflow metabolism. The strain VH34 has an additional deletion of the pyruvate kinase A gene (pykA) to increase pDNA formation. pDNA formation rates as well as kinetic and stoichiometric parameters were investigated in dependence of the growth rate within a range from 0.02 to 0.25 h(-1). Differences between strains were found in terms of the biomass yields on nitrogen and oxygen, as well as on the cell maintenance coefficients. The deletion of pykA led to a significantly increased pDNA yield and productivity. At an optimal growth rate of 0.20 h(-1) it was nearly 60% higher than that of W3110 and VH33. Metabolic fluxes calculated by metabolite balance analysis showed differences mainly in reactions catalyzed by pyruvate kinase and glucose 6-phosphate dehydrogenase. The obtained data are useful for the design of cultivation schemes for pDNA production by E. coli.
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret