MilliporeSigma
  • [Antineutrophil cytoplasmic antibody (Anca)-associated autoimmune disease induced by propylthiouracil].
[Antineutrophil cytoplasmic antibody (Anca)-associated autoimmune disease induced by propylthiouracil].
Arquivos brasileiros de endocrinologia e metabologia (2007-04-17)
Nádila Cecyn Pietszkowski, Gisah Amaral de Carvalho, Hevelyn Noemberg de Souza, Cláudio Rogério Werka, Luís Augusto Borazo, Hans Graf, Mauricio de Carvalho
ABSTRACT
Antithyroid drugs sometimes cause severe complications. Propylthiouracil (PTU) can be associated to ANCA positive vasculitis, most often related to myeloperoxidase subtype (ANCA-MPO). Our objective is to describe a female patient with Graves' disease, who developed PTU induced-autoimmune disease, with cutaneous, pulmonary, and renal lesions, associated with ANCA. Histopathological examination revealed diffuse pulmonary hemorrhage, and focal segmental glomerulosclerosis at the kidney biopsy. She was treated with systemic corticosteroid therapy and cyclophosphamide, with clinical improvement. This case highlights the need for greater awareness of this relatively rare adverse effect of propylthiouracil.
Product Description
Propylthiouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
6-Propyl-2-thiouracil, VETRANAL®, analytical standard
Sigma-Aldrich
6-Propyl-2-thiouracil, enzyme inhibitor