MilliporeSigma
  • Kinetics and metabolism of the Fusarium toxin deoxynivalenol in farm animals: consequences for diagnosis of exposure and intoxication and carry over.
Kinetics and metabolism of the Fusarium toxin deoxynivalenol in farm animals: consequences for diagnosis of exposure and intoxication and carry over.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2013-07-23)
Sven Dänicke, Ulrike Brezina
ABSTRACT
The knowledge of factors influencing the kinetics, metabolism and bioavailability of the Fusarium toxin deoxynivalenol (DON) is a basic prerequisite for evaluation of the transfer (carry over) of the toxin and its metabolites into edible tissues, and for a physiological specimen-based diagnosis of intoxication. These aspects are addressed in the present review, and potentials and pitfalls of the suitability of analysis of physiological samples for evaluation of the DON exposure as a veterinary tool are discussed. For example, the farm animal species was shown to be a determining factor influencing the metabolic profile and the bioavailability of DON. Although linear relationships were derived between DON exposure of ad libitum and restrictively fed animals and DON or de-epoxy-DON concentration in the blood of pigs, dairy cows and sheep, it has to be considered that individual values might markedly deviate from these relationships, which makes interpretation of measured concentrations of DON and its metabolites difficult. The situation is further complicated by the lack of established relationships between DON residues in physiological matrices and the adverse effects of DON on the health and performance of farm animals.
Product Description
Supelco
Deoxynivalenol solution, 100 μg/mL in acetonitrile, analytical standard
4-Deoxynivalenol in acetonitrile, IRMM®, certified reference material
Sigma-Aldrich
Deoxynivalenol