MilliporeSigma
  • Arylethynylacridines: electrochemiluminescence and photophysical properties.
Arylethynylacridines: electrochemiluminescence and photophysical properties.
Organic & biomolecular chemistry (2004-10-27)
Arumugasamy Elangovan, Hsing-Hua Chiu, Shu-Wen Yang, Tong-Ing Ho
ABSTRACT
Electrogenerated chemiluminescence (ECL) of six new ethyne-based acridine derivatives (1-6) has been studied. The new acridine derivatives were synthesized by cross-coupling of 9-chloroacridine and corresponding donor-substituted phenylethynes under modified Sonogashira conditions. The donor groups were varied in the order of increasing steric hindrance and donor strength at the donor site. The solution phase photophysical properties and ECL of these compounds were studied comparatively in acetonitrile solvent. The UV-Visible spectra of compounds 1-5 exhibit closely the same maxima. Density functional theory (DFT) has been invoked to analyze and understand the unexpected UV-Visible absorption behavior. Compounds with weak electron donors produce excimer ECL irrespective of steric hindrance at the donor site, while the compound with a stronger donor gives rise to ECL that is blue-shifted with respect to its photoluminescence spectrum. All except one of these compounds also exhibit solid state fluorescence which may be useful for solid state devices such as organic light emitting diodes (OLEDs) and as laser dyes. The observed properties are discussed with reference to the structure of the compounds synthesized.
Sigma-Aldrich
9-Chloroacridine, 97%
Pricing and availability is not currently available.