MilliporeSigma
  • Direct sulfonation of methane to methanesulfonic acid by sulfur trioxide catalyzed by cerium (IV) sulfate in the presence of molecular oxygen.
Direct sulfonation of methane to methanesulfonic acid by sulfur trioxide catalyzed by cerium (IV) sulfate in the presence of molecular oxygen.
Advanced Synthesis & Catalysis (2004)
Mukhopadhyay S and Bell AT.