MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • METEORIN-LIKE is a cytokine associated with barrier tissues and alternatively activated macrophages.
METEORIN-LIKE is a cytokine associated with barrier tissues and alternatively activated macrophages.
Clinical immunology (Orlando, Fla.) (2014-12-09)
Irina Ushach, Amanda M Burkhardt, Cynthia Martinez, Peter A Hevezi, Peter Arne Gerber, Bettina Alexandra Buhren, Holger Schrumpf, Ricardo Valle-Rios, Monica I Vazquez, Bernhard Homey, Albert Zlotnik
ABSTRACT
Cytokines are involved in many functions of the immune system including initiating, amplifying and resolving immune responses. Through bioinformatics analyses of a comprehensive database of gene expression (BIGE: Body Index of Gene Expression) we observed that a small secreted protein encoded by a poorly characterized gene called meteorin-like (METRNL), is highly expressed in mucosal tissues, skin and activated macrophages. Further studies indicate that Metrnl is produced by Alternatively Activated Macrophages (AAM) and M-CSF cultured bone marrow macrophages (M2-like macrophages). In the skin, METRNL is expressed by resting fibroblasts and IFNγ-treated keratinocytes. A screen of human skin-associated diseases showed significant over-expression of METRNL in psoriasis, prurigo nodularis, actinic keratosis and atopic dermatitis. METRNL is also up-regulated in synovial membranes of human rheumatoid arthritis. Taken together, these results indicate that Metrnl represents a novel cytokine, which is likely involved in both innate and acquired immune responses.
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG