MilliporeSigma
  • Activation of the leukemia plasmacytoid dendritic cell line PMDC05 by Toho-1, a novel IDO inhibitor.
Activation of the leukemia plasmacytoid dendritic cell line PMDC05 by Toho-1, a novel IDO inhibitor.
Anticancer research (2014-07-31)
Akie Yamahira, Miwako Narita, Minami Iwabuchi, Takayoshi Uchiyama, Shunpei Iwaya, Rie Ohiwa, Yoshinori Nishizawa, Takafumi Suzuki, Yusaku Yokoyama, Shigeo Hashimoto, Jun Takizawa, Hirohito Sone, Masuhiro Takahashi
ABSTRACT
Indoleamine-2,3-dioxygenase (IDO) is a rate-limiting enzyme for tryptophan metabolism and plays an immunosuppressive role. Antigen-presenting cells, when activated, increase the expression of IDO, which results in the suppression of subsequent immune reaction. A novel IDO inhibitor, Toho-1, was explored for its applicability to immunotherapy. We investigated the effects of Toho-1 on antigen presentation and antigen-specific cytotoxic T-lymphocyte-inducing ability of leukemia plasmacytoid dendritic cell line PMDC05, which was established in our laboratory. While antigen presentation-associated molecules in PMDC05 cells were increased by stimulation with lipopolysaccharide and interferon-γ, IDO mRNA and protein expression were also enhanced. Such treatment of PMDC05 cells in combination with Toho-1 enhanced the antigen-presenting and CTL-inducing ability of PMDC05 cells. These findings suggest the ability of Toho-1 to potentiate antigen-presenting cells and its applicability in immunotherapy of cancer.