MilliporeSigma
  • Comparative metabolomics analysis on hematopoietic functions of herb pair Gui-Xiong by ultra-high-performance liquid chromatography coupled to quadrupole time-of-flight mass spectrometry and pattern recognition approach.
Comparative metabolomics analysis on hematopoietic functions of herb pair Gui-Xiong by ultra-high-performance liquid chromatography coupled to quadrupole time-of-flight mass spectrometry and pattern recognition approach.
Journal of chromatography. A (2014-05-06)
Weixia Li, Yuping Tang, Jianming Guo, Erxin Shang, Yefei Qian, Linyan Wang, Li Zhang, Pei Liu, Shulan Su, Dawei Qian, Jin-ao Duan
ABSTRACT
The compatibility of Angelicae Sinensis Radix (Danggui, DG) and Chuanxiong Rhizoma (Chuanxiong, CX), a famous herb pair Gui-Xiong (GX), can produce synergistic and complementary hematopoiesis. In present study, global metabolic profiling with ultra-high-performance liquid chromatography coupled to quadrupole time-of-flight mass spectrometry (UHPLC-QTOF/MS) combined with pattern recognition method was performed to discover the underlying hematopoietic regulation mechanisms of DG, CX and GX on hemolytic and aplastic anemia rats (HAA) induced by acetyl phenylhydrazine (APH) and cyclophosphamide (CP). Thirteen endogenous metabolites contributing to the separation of model group and control group were tentatively identified. The levels of LPCs including lysoPC (18:0), lysoPC (20:4), lysoPC (16:0) and lysoPC (18:2), sphinganine, nicotinic acid, thiamine pyrophosphate, phytosphingosine, and glycerophosphocholine increased significantly (p<0.05) in HAA, while the levels of oleic acid, 8,11,14-eicosatrienoic acid, ceramides (d18:1/14:0), and 17a-hydroxypregnenolone decreased significantly (p<0.05) in comparison with control rats. Those endogenous metabolites were chiefly involved in thiamine metabolism and sphingolipid metabolism. The metabolic deviations could be regulated closer to normal level after DG, CX and GX intervention. In term of hematopoietic function, GX was the most effective as shown by the relative distance in PLS-DA score plots and relative intensity of metabolomic strategy, reflecting the synergic action between DG and CX. The relative distance calculation was firstly used in metabolomics for semi-quantization.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%