MilliporeSigma
  • Linifanib (ABT-869), enhances cytotoxicity with poly (ADP-ribose) polymerase inhibitor, veliparib (ABT-888), in head and neck carcinoma cells.
Linifanib (ABT-869), enhances cytotoxicity with poly (ADP-ribose) polymerase inhibitor, veliparib (ABT-888), in head and neck carcinoma cells.
Oral oncology (2014-04-17)
Heng-Wei Hsu, Rosalia de Necochea-Campion, Vonetta Williams, Penelope J Duerksen-Hughes, Alfred A Simental, Robert L Ferris, Chien-Shing Chen, Saied Mirshahidi
ABSTRACT
PARP inhibitors (PARPi) may provide an opportunity to gain selective killing of tumor cells which have deficiencies in cellular DNA repair systems. We previously demonstrated linifanib (ABT-869), a multi-receptor tyrosine kinase inhibitor of VEGF and PDGF receptor families, radiosensitized Head and Neck Squamous Cell Carcinoma cells (HNSCC) via inhibiting STAT3 activation. Given that STAT3 can modulate DNA damage response (DDR) pathway, in this study, we evaluate the effects of linifanib to enhance cytotoxicity with the PARPi, veliparib (ABT-888), in HNSCC. UMSCC-22A and UMSCC-22B cells were treated with linifanib (ABT-869) and veliparib (ABT-888). Cell viability, cytotoxicity, apoptosis induction, DNA single strand break (SSB) and double strand break (DSB) damages were examined by MTT assay, colony formation assay, flow cytometry and comet assay. In addition, the expression of DNA homologous recombination repair protein Rad51, γH2AX, a double strand break marker and cleaved PARP, an apoptotic cell death marker, were assessed using western immunoblotting. Combination treatment resulted in more cell growth inhibition, induction of apoptosis, DNA damages and double strand breaks, lower expression of Rad51, increase γH2AX expression and PARP cleavage. These data suggest the possibility of combining targeted therapeutic such as linifanib with veliparib to augment the inhibition of cell growth and apoptosis via synthetic lethality in HNSCC cells. Thus, it may provide a novel therapeutic strategy and improve efficacy and outcome in HNSCC.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide solution