MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Development, optimization and validation of a multimethod for the determination of 36 mycotoxins in wines by liquid chromatography-tandem mass spectrometry.
Development, optimization and validation of a multimethod for the determination of 36 mycotoxins in wines by liquid chromatography-tandem mass spectrometry.
Talanta (2014-08-17)
Ionara R Pizzutti, Andre de Kok, Jos Scholten, Laís W Righi, Carmem D Cardoso, Graciele Necchi Rohers, Rosselei C da Silva
ABSTRACT
A fast and efficient multimethod for the determination of 36 mycotoxins in wine, using ultra-performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS), was developed, optimized, validated and implemented in routine analysis. A simplified, quick extraction was performed with acetonitrile, derived from the QuEChERS (quick, easy, cheap, effective, rugged and safe) approach, which was traditionally developed for pesticides analysis. This study aimed at a single extraction and chromatographic separation for 36 mycotoxins. Optimization tests were performed to find the proper ratio of wine: water and extraction solvent and the need for an additional buffering step with ammonium formate/formic acid and a dispersive SPE cleanup with various sorbents. The dSPE steps did not show significant improvement in analysis results, therefore, it was not applied in the final method to be validated. The mycotoxins were separated and detected on a UPLC-MS/MS system, used in the ESI positive ionization mode. The various mycotoxins were divided in three different concentration level groups, according to their sensitivity in UPLC-MS/MS. The validation was performed by analyzing recovery samples at three different spike levels with six replicates (n=6) at each level. Linearity (r(2)) of calibration curves, accuracy (recovery %), instrument limits of detection and method limits of quantification (LOD and LOQ), precision (RSD%) and matrix effects (%) were determined for each individual mycotoxin. From the 36 mycotoxins analyzed by UPLC-MS/MS (ESI+), 35 showed average recoveries in the range 70-120%, and 86% of these with a RSD≤20% at the lowest spike level (for Group I, II and III, respectively, 1, 50 and 10 µg kg(-1)). The higher spike levels showed even better results. Only nivalenol could not be quantified at any concentration level. The method LOQ for 86% of the mycotoxins studied was the lowest spike level tested. The matrix effect observed was low for most mycotoxins analyzed and had no significant influence on the analytical results obtained. The developed procedure was applied successfully in routine analysis in a survey of wine samples originating from different countries.
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG