MilliporeSigma
  • Identification of phenanthrene derivatives in Aerides rosea (Orchidaceae) using the combined systems HPLC-ESI-HRMS/MS and HPLC-DAD-MS-SPE-UV-NMR.
Identification of phenanthrene derivatives in Aerides rosea (Orchidaceae) using the combined systems HPLC-ESI-HRMS/MS and HPLC-DAD-MS-SPE-UV-NMR.
Phytochemical analysis : PCA (2014-08-19)
Veronika Cakova, Aurélie Urbain, Cyril Antheaume, Nicole Rimlinger, Patrick Wehrung, Frédéric Bonté, Annelise Lobstein
ABSTRACT
In our continued efforts to contribute to the general knowledge on the chemical diversity of orchids, we have decided to focus our investigations on the Aeridinae subtribe. Following our previous phytochemical study of Vanda coerulea, which has led to the identification of phenanthrene derivatives, a closely related species, Aerides rosea Lodd. ex Lindl. & Paxton, was chosen for investigation. To identify new secondary metabolites, and to avoid isolation of those already known, by means of the combined systems HPLC-DAD(diode-array detector) with high-resolution tandem mass spectrometry (HRMS/MS) and HPLC-DAD-MS-SPE(solid-phase extraction)-UV-NMR. A dereplication strategy was developed using a HPLC-DAD-HRMS/MS targeted method and applied to fractions from A. rosea stem extract. Characterisation of unknown minor compounds was then performed using the combined HPLC-DAD-MS-SPE-UV-NMR system. The dereplication method allowed the characterisation of four compounds (gigantol, imbricatin, methoxycoelonin and coelonin), previously isolated from Vanda coerulea stem extract. The analyses of two fractions permitted the identification of five additional minor constituents including one phenanthropyran, two phenanthrene and two dihydrophenanthrene derivatives. The full set of NMR data of each compound was obtained from microgram quantities. Nine secondary metabolites were characterised in A. rosea stems, utilising HPLC systems combined with high-resolution analytical systems. Two of them are newly described phenanthrene derivatives: aerosanthrene (5-methoxyphenanthrene-2,3,7-triol) and aerosin (3-methoxy-9,10-dihydro-2,5,7-phenanthrenetriol).
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%