MilliporeSigma
  • Dihydropyrimidinase-like 3 is a putative hepatocellular carcinoma tumor suppressor.
Dihydropyrimidinase-like 3 is a putative hepatocellular carcinoma tumor suppressor.
Journal of gastroenterology (2014-09-01)
Hisaharu Oya, Mitsuro Kanda, Hiroyuki Sugimoto, Dai Shimizu, Hideki Takami, Soki Hibino, Ryoji Hashimoto, Yukiyasu Okamura, Suguru Yamada, Tsutomu Fujii, Goro Nakayama, Masahiko Koike, Shuji Nomoto, Michitaka Fujiwara, Yasuhiro Kodera
ABSTRACT
Patients with hepatocellular carcinoma (HCC) may relapse after curative resection. Sensitive biomarkers for HCC are required to enhance disease management. Dihydropyrimidinase-like 3 (DPYSL3) suppresses cell proliferation and tumorigenicity of certain malignancies; however, its role in HCC is unknown. The expression levels of DPYSL3 and genes encoding potential interacting proteins vascular endothelial growth factor (VEGF), focal adhesion kinase (FAK), ezrin, and cellular src were determined using RT-PCR. Further, we determined the methylation status of the DPYSL3 promoter in HCC cells lines and the effect of inhibiting DPYSL3 expression on their phenotype. DPYSL3 expression was determined in 151 pairs of resected liver tissues. DPYSL3 mRNA levels were down-regulated in most HCC cell lines with DPYSL3 promoter hypermethylation, and expression was restored after demethylation. DPYSL3 expression levels inversely correlated with those of VEGF and FAK. Knockdown of DPYSL3 significantly increased migration and the invasive properties of HCC cells. The mean level of DPYSL3 mRNA was significantly lower in HCC tissues compared with corresponding noncancerous tissues. The expression patterns of DPYSL3 mRNA and protein were consistent. DPYSL3 mRNA expression in HCC tissues inversely correlated with preoperative serum tumor markers and was significantly lower in patients with extrahepatic recurrences. Disease-specific and recurrence-free survival was significantly shorter in patients with down-regulated DPYSL3 expression. Our results indicate that DPYSL3 is a putative HCC tumor suppressor, and promoter hypermethylation potently regulates DPYSL3 transcription. Down-regulation of DPYSL3 expression in HCC tissues may serve as a predictive biomarker for HCC after curative resection.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dpysl3
Sigma-Aldrich
MISSION® esiRNA, targeting human DPYSL3