MilliporeSigma
  • The Effects of Cadmium at Low Environmental Concentrations on THP-1 Macrophage Apoptosis.
The Effects of Cadmium at Low Environmental Concentrations on THP-1 Macrophage Apoptosis.
International journal of molecular sciences (2015-09-16)
Tomasz Olszowski, Irena Baranowska-Bosiacka, Izabela Gutowska, Katarzyna Piotrowska, Katarzyna Mierzejewska, Jan Korbecki, Mateusz Kurzawski, Maciej Tarnowski, Dariusz Chlubek
ABSTRACT
Cadmium at environmental concentrations is a risk factor for many diseases, including cardiovascular and neurodegenerative diseases, in which macrophages play an important role. The aim of this study was to evaluate the effects of cadmium at low environmental (nanomolar) concentrations on apoptotic processes in THP-1(acute monocytic leukemia cells line)-derived macrophages, with special focus on mitochondrial events involved. Macrophages were incubated with various cadmium chloride (CdCl₂) solutions for 48 h at final concentrations of 5 nM, 20 nM, 200 nM and 2 µM CdCl₂. Cell viability was measured using flow cytometry. Flow cytometric measurement (annexin V/FITC (annexin V/fluorescein isothiocyanate) and PI (propidium iodide) double staining) was used to quantify the extent of apoptosis. Fluorescence and confocal microscopy were used for imaging of apoptosis process. Changes in mitochondrial membrane potential were monitored using cytofluorimetry after cell staining with JC-1(5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazol-carbocyane iodide) probe. Mitochondrial ROS (reactive oxygen species) levels were measured cytofluorimetrically after incubation of cells with mitochondrial superoxide indicator (MitoSOX) red fluorescent marker. The mRNA expression of Bcl-2 and Bax was analysed with qRT-PCR. Our study demonstrates that cadmium, even at low environmental concentrations, exerts mitochondrial toxicity in THP-1 macrophages. Forty-eight-hour exposure to very low concentrations reduces cell viability and results in cell death by apoptosis and necrosis. The decrease in mitochondrial membrane potential, increased ROS production, increased Bax and decreased Bcl-2 mRNA expression are mitochondrial events involved in cadmium-induced apoptosis.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Cadmium chloride
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)