MilliporeSigma
Recycling of ladle slag in cement composites: Environmental impacts.
Waste management (New York, N.Y.) (2015-05-27)
Vesna Zalar Serjun, Ana Mladenovič, Breda Mirtič, Anton Meden, Janez Ščančar, Radmila Milačič
ABSTRACT
In the present work compact and ground cement composites in which 30% of cement by mass was replaced by ladle slag were investigated for their chemical and physico-mechanical properties. To evaluate long-term environmental impacts, leachability test based on diffusion, which combined both, diffusion and dissolution of contaminants, was performed in water and saline water. Total element concentrations and Cr(VI) were determined in leachates over a time period of 180days. At the end of the experiment, the mineralogical composition and the physico-mechanical stability of cement composites was also assessed. The results revealed that Cr(III) and Cr(VI) were immobilized by the hydration products formed in the cement composites with the addition of ladle slag. Cr(VI) content originating from the cement was also appreciably reduced by Fe(II) from minerals present in the added ladle slag, which thus had significant positive environmental effects. Among metals, only Mo and Ba were leached in elevated concentrations, but solely in ground cement composites with the addition of ladle slag. Lower V concentrations were observed in leachates of ground than compact composite. It was demonstrated that the presence of ladle slag in cement composites can even contribute to improved mortar resistance. The investigated ladle slag can be successfully implemented in cement composites as supplementary cementitious material.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis