MilliporeSigma
  • Prioritization and association analysis of murine-derived candidate genes in anxiety-spectrum disorders.
Prioritization and association analysis of murine-derived candidate genes in anxiety-spectrum disorders.
Biological psychiatry (2011-08-30)
John M Hettema, Bradley T Webb, An-Yuan Guo, Zhongming Zhao, Brion S Maher, Xiangning Chen, Seon-Sook An, Cuie Sun, Steven H Aggen, Kenneth S Kendler, Po-Hsiu Kuo, Takeshi Otowa, Jonathan Flint, Edwin J van den Oord
ABSTRACT
Anxiety disorders are common psychiatric conditions that are highly comorbid with each other and related phenotypes such as depression, likely due to a shared genetic basis. Fear-related behaviors in mice have long been investigated as potential models of anxiety disorders, making integration of information from both murine and human genetic data a powerful strategy for identifying potential susceptibility genes for these conditions. We combined genome-wide association analysis of fear-related behaviors with strain distribution pattern analysis in heterogeneous stock mice to identify a preliminary list of 52 novel candidate genes. We ranked these according to three complementary sources of prior anxiety-related genetic data: 1) extant linkage and knockout studies in mice, 2) a meta-analysis of human linkage scans, and 3) a preliminary human genome-wide association study. We genotyped tagging single nucleotide polymorphisms covering the nine top-ranked regions in a two-stage association study of 1316 subjects from the Virginia Adult Twin Study of Psychiatric and Substance Use Disorders chosen for high or low genetic loading for anxiety-spectrum phenotypes (anxiety disorders, neuroticism, and major depression). Multiple single nucleotide polymorphisms in the PPARGC1A gene demonstrated association in both stages that survived gene-wise correction for multiple testing. Integration of genetic data across human and murine studies suggests PPARGC1A as a potential susceptibility gene for anxiety-related disorders.