- Limited effect of ozone reductions on the 20-year photosynthesis trend at Harvard forest.
Limited effect of ozone reductions on the 20-year photosynthesis trend at Harvard forest.
Global change biology (2016-10-21)
Xu Yue, Trevor F Keenan, William Munger, Nadine Unger
PMID27028880
ABSTRACT
Ozone (O
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)